Learn More
Perfluorohexadecanoic acid 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Specifications
Specifications
| Quantity | 1 mL |
| Concentration | 50 μg/mL |
| Certifications/Compliance | ISO/IEC 17025 |
| Solvent or Matrix | Methanol/Water |