missing translation for 'onlineSavingsMsg'
Learn More

Perfluorohexanesulfonamide (>75%), TRC

Product Code. 30009395
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009395 500 mg 500mg
17911025 100 mg 100mg
17994295 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009395 Supplier Toronto Research Chemicals Supplier No. TRCP999355500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41997-13-1
Molecular Formula C6 H2 F13 N O2 S
Molecular Weight (g/mol) 399.1298
Synonym Perfluorohexane Sulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluoro-1-hexanesulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluorohexan-1-sulfonamide, AI 3-10702, FHxSA, Perfluorohexanesulfonamide, Perfluorohexylsulfonamide, n-Perfluorohexylsulfonamide, FHxSA, 1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluoro-1-hexanesulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluorohexan-1-sulfonamide, AI 3-10702, FHxSA, Perfluorohexanesulfonamide, Perfluorohexylsulfonamide, n-Perfluorohexylsulfonamide, PFHxSA
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane-1-sulfonamide
SMILES NS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H2F13NO2S/c7-1(8,3(11,12)5(15,16)17)2(9,10)4(13,14)6(18,19)23(20,21)22/h(H2,20,21,22)
Formula Weight 398.9599
Chemical Name or Material Perfluorohexane Sulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.