missing translation for 'onlineSavingsMsg'
Learn More

Perfluorohexanoic Acid (>90%), TRC

Product Code. 17920735
Change view
Click to view available options
Quantity:
2.5 g
10 g
25 g
Unit Size:
10g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17920735 2.5 g 2.5g
17984575 25 g 25g
17913355 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17920735 Supplier Toronto Research Chemicals Supplier No. TRCP2865252.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307-24-4
Formula Weight 313.9801
InChI Formula InChI=1S/C6HF11O2/c7-2(8,1(18)19)3(9,10)4(11,12)5(13,14)6(15,16)17/h(H,18,19)
IUPAC Name 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexanoic acid
Molecular Formula C6 H F11 O2
Molecular Weight (g/mol) 314.05
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Synonym Hexanoic acid, undecafluoro- (6CI,7CI,8CI,9CI), 2,2,3,3,4,4,5,5,6,6,6-Undecafluorohexanoic acid, NSC 5213, PFHxA, Perfluoro-1-pentanecarboxylic acid, Perfluorohexanoic acid
Chemical Name or Material Perfluorohexanoic acid

Specifications

Quantity 2.5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.