Learn More
Perfluorononan-1-oic-acid Ammonium Salts, TRC
Product Code. 18082040
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18082040 1 g 1g
18060650 500 mg 500mg
18014050 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18082040 Supplier Toronto Research Chemicals Supplier No. TRCP2865551G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4149-60-4
Molecular Formula C9 F17 O2 . H4 N
Molecular Weight (g/mol) 481.106
Synonym Heptadecafluoro-nonanoic Acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-Heptadecafluorononanoic Acid, C 1800, Heptadecafluorononanoic Acid, PFNA, Perfluorononanoic Acid, Perfluoropelargonic Acid, Nonanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro-, ammonium salt (1:1), Nonanoic acid, heptadecafluoro-, ammonium, salt (8CI,9CI), Ammonium perfluorononanoate, Ammonium perfluoropelargonate
IUPAC Name ammonium;2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoate
SMILES [NH4+].[O-]C(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifications
Quantity 1 g
InChI Formula InChI=1S/C9HF17O2.H3N/c10-2(11,1(27)28)3(12,13)4(14,15)5(16,17)6(18,19)7(20,21)8(22,23)9(24,25)26;/h(H,27,28);1H3
Formula Weight 480.9971
Percent Purity >95%
Chemical Name or Material Perfluorononan-1-oic-acid Ammonium Salts
Grade HPLC