Learn More
Perfluorononanoic acid, 97%
Product Code. 11419070
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
missing translation for 'unitSize'
100g
25g
5g
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11419070
missing translation for 'mfr': Thermo Scientific Alfa Aesar A13980.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
CAS: 375-95-1 | C9HF17O2 | 464.078 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 375-95-1
Molecular Formula C9HF17O2
Molecular Weight (g/mol) 464.078
MDL Number MFCD00039605
InChI Key UZUFPBIDKMEQEQ-UHFFFAOYSA-N
Synonym perfluorononanoic acid, heptadecafluorononanoic acid, perfluoro-n-nonanoic acid, nonanoic acid, heptadecafluoro, perfluorononan-1-oic acid, pfna, heptadecafluoropelargonic acid, perfluornonansaeure, heptadecafluornonansaeure, heptadekafluornonansaeure
PubChem CID 67821
ChEBI CHEBI:38397
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoic acid
SMILES C(=O)(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
Specifications
Melting Point 63°C to 68°C
Boiling Point 218°C
Flash Point None
Linear Formula CF3(CF2)7CO2H
Quantity 100 g
UN Number UN3261
Beilstein 1897287
Formula Weight 464.07
Percent Purity 97%
Chemical Name or Material Perfluorononanoic acid