Learn More
BINDER™ Shelf, Perforated
Product Code. 16684173 Shop All BINDER Products
Click to view available options
Dimensions (L x W x H):
620 x 510 x 50 mm
Unit Size:
Each
Product Code. 16684173
Brand: BINDER™ 80121996
Only null left
Add to basket
Add to basket
Perforated Shelf is stainless steel.
  • Shelf
  • Perforated
  • Made of stainless steel
Specifications
Description Shelf, perforated - Stainless steel, divided, for divided inner door
For Use With (Equipment) BINDER Chambers CB260, CB-S260
Material Stainless Steel
Type Divided Shelf
Model CB 260 (E7)
Dimensions (L x W x H) 620 x 510 x 50 mm
Length (English) 2.03 ft.
Length (Metric) 620 mm
Width (English) 1.67 ft.
Width (Metric) 510 mm
Height (English) 1.97 in.
Height (Metric) 50 mm
Shipping Weight (Metric) 4.6 Kg
Weight (English) 9.26 lb.
Weight (Metric) 4.2 kg