Learn More
Perforin-1 Monoclonal Antibody (OTI3D7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA814190S
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
Perforin-1 Monoclonal antibody specifically detects Perforin-1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Perforin-1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P14222 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 5551 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| OTI3D7 | |
| Unconjugated | |
| PRF1 | |
| Cyta; Cytolysin; FLH2; HPLH2; lymphocyte pore forming protein; lymphocyte pore-forming protein; OMAK; OTTHUMP00000019759; P1; perforin 1; perforin 1 (pore forming protein); Perforin1; perforin-1; Pfn; PFN1; Pfp; PGFL; PIGF; PIGF-2; PLGF; pore forming protein; pore-forming protein; PRF1; Prf-1; PRF1 (pore forming protein 1); RATCYTA; RP11-710A11.3 | |
| Human recombinant protein fragment corresponding to amino acids 351-555 of human Perforin-1 (NP_005032) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.