missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perforin Antibody (1031751), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB103853W
This item is not returnable.
View return policy
Description
Perforin Monoclonal antibody specifically detects Perforin in Human samples. It is validated for Flow Cytometry
Specifications
| Perforin | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1031751 | |
| Intracellular Staining by Flow Cytometry | |
| Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin | |
| Chinese Hamster Ovary cell line CHO-derived human Perforin, Pro22-Trp555, Accession # P14222 | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Immunology, Innate Immunity | |
| 5551 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction