missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perforin Antibody (CE2.10), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97440V
This item is not returnable.
View return policy
Description
Perforin Monoclonal specifically detects Perforin in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Perforin | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PRF1 | |
| Recombinant mouse perforin (aa 98-534). | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Immunology, Innate Immunity | |
| Recognizes an epitope in the region aa 402-534 of human and mouse perforin. Does NOT detect Human in Western Blot. | |
| Store at 4°C in the dark. |
| Western Blot | |
| CE2.10 | |
| Western Blot | |
| Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction