Learn More
Pergafast 201, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Pergafast 201 |
| CAS | 232938-43-1 |
| Analyte or Component Names | Pergafast 201 |
| Shipping Condition | +20°C |
| Molecular Formula | C21 H20 N2 O6 S2 |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | 4-Methyl-N-[[[3-[[(4-methylphenyl)sulfonyl]oxy]phenyl]amino]carbonyl]benzenesulfonamide, DP 201, N-(4-Methylphenylsulfonyl)-N'-[3-(4-methylphenylsulfonyloxy)phenyl]urea, N-(p-Toluenesulfonyl)-N'-(3-p-toluenesulfonyloxyphenyl)urea, N-p-Tolylsulfonyl-N'-3-(p-tolylsulfonyloxy)phenylurea, PF 201, Pergafast 201 |
| InChI Formula | InChI=1S/C21H20N2O6S2/c1-15-6-10-19(11-7-15)30(25,26)23-21(24)22-17-4-3-5-18(14-17)29-31(27,28)20-12-8-16(2)9-13-20/h3-14H,1-2H3,(H2,22,23,24) |
| SMILES | Cc1ccc(cc1)S(=O)(=O)NC(=O)Nc2cccc(OS(=O)(=O)c3ccc(C)cc3)c2 |
| IUPAC Name | [3-[(4-methylphenyl)sulfonylcarbamoylamino]phenyl] 4-methylbenzenesulfonate |
| Show More |