Learn More
Pergafast 201, TRC
Product Code. 17984015
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17984015 1 g 1g
17963765 500 mg 500mg
17951875 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17984015 Supplier Toronto Research Chemicals Supplier No. TRCP2871801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 232938-43-1
Molecular Formula C21 H20 N2 O6 S2
Molecular Weight (g/mol) 460.52
Synonym 4-Methyl-N-[[[3-[[(4-methylphenyl)sulfonyl]oxy]phenyl]amino]carbonyl]benzenesulfonamide, DP 201, N-(4-Methylphenylsulfonyl)-N'-[3-(4-methylphenylsulfonyloxy)phenyl]urea, N-(p-Toluenesulfonyl)-N'-(3-p-toluenesulfonyloxyphenyl)urea, N-p-Tolylsulfonyl-N'-3-(p-tolylsulfonyloxy)phenylurea, PF 201, Pergafast 201
IUPAC Name [3-[(4-methylphenyl)sulfonylcarbamoylamino]phenyl] 4-methylbenzenesulfonate
SMILES Cc1ccc(cc1)S(=O)(=O)NC(=O)Nc2cccc(OS(=O)(=O)c3ccc(C)cc3)c2
Specifications
Quantity 1 g
InChI Formula InChI=1S/C21H20N2O6S2/c1-15-6-10-19(11-7-15)30(25,26)23-21(24)22-17-4-3-5-18(14-17)29-31(27,28)20-12-8-16(2)9-13-20/h3-14H,1-2H3,(H2,22,23,24)
Formula Weight 460.0763
Chemical Name or Material Pergafast 201