missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perilipin-2/ADFP Antibody (ADFP/1365), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54369PCP
This item is not returnable.
View return policy
Description
Perilipin-2/ADFP Monoclonal antibody specifically detects Perilipin-2/ADFP in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Protein Array, CyTOF
Specifications
| Perilipin-2/ADFP | |
| Monoclonal | |
| PerCP | |
| ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2 | |
| Recombinant fragment (around aa 249-376) of human Perilipin-2/ADFP protein (exact sequence is proprietary) (Uniprot: Q99541) | |
| 100 μg | |
| Cancer, Cardiovascular Biology, Diabetes Research, Epigenetics, Hypoxia, Lipid and Metabolism, Lipid Droplets, metabolism, Signal Transduction | |
| 123 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Protein Array, CyTOF | |
| ADFP/1365 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction