missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Periostin Monoclonal Antibody (OTI2D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803713S
This item is not returnable.
View return policy
Description
Periostin Monoclonal antibody specifically detects Periostin in Human samples. It is validated for Western BlotSpecifications
| Periostin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15063 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10631 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2D10 | |
| Unconjugated | |
| Postn | |
| A630052E07Rik; AI747096; fasciclin I-like; Fasciclin-I like; LOW QUALITY PROTEIN: periostin; MGC119510; MGC119511; OSF 2; Osf2; OSF-2; osteoblast specific factor 2; osteoblast specific factor 2 (fasciclin I-like); osteoblast-specific factor 2; PDLPOSTN; peri; periodontal ligament-specific periostin; periostin; periostin variant 1; periostin variant 2; periostin variant 3; periostin variant 4; periostin variant 5; periostin variant 6; periostin variant 7; periostin variant 8; periostin variant 9; periostin, osteoblast specific factor; periostin-like factor; Plf; PN; POSTN; POSTN protein; RP11 412K4.1; RP11-412K4.1 | |
| Human recombinant protein fragment corresponding to amino acids 538-781 of human POSTN produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction