missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST2 Mouse anti-Human, Clone: OTI21A7, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810124S
This item is not returnable.
View return policy
Description
ST2 Monoclonal antibody specifically detects ST2 in Human samples. It is validated for ELISASpecifications
| ST2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q01638 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 9173 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| OTI21A7 | |
| Unconjugated | |
| IL1RL1 | |
| DER4; Fit1; Fit-1; Fos-responsive gene 1 protein; growth stimulation-expressed; homolog of mouse growth stimulation-expressed; IL-1 R4; IL1RL1; IL-1RL1; IL33R; IL-33R; interleukin 1 receptor like 1; interleukin 1 receptor-like 1; interleukin 1 receptor-related protein; Interleukin1 receptor like 1; interleukin-1 receptor-like 1; Interleukin-33 receptor alpha chain; Ly84; lymphocyte antigen 84; Protein ST2; Protein T1; sIL 33R; soluble IL 33 Receptor; soluble IL 33R; St2; ST2L; St2-rs1; ST2V; Ste2; T1; T1/ST2 | |
| FulllengthhumanrecombinantproteinofhumanST2(NP_057316)producedinHEK293E | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction