missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Periostin Monoclonal Antibody (OTI6A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803701
This item is not returnable.
View return policy
Description
Enhances incorporation of BMP1 in the fibronectin matrix of connective tissues, and subsequent proteolytic activation of lysyl oxidase LOX. Induces cell attachment and spreading and plays a role in cell adhesion. May play a role in extracellular matrix mineralization.Specifications
| Periostin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15063 | |
| POSTN | |
| Human recombit protein fragment corresponding to amino acids 538-781 of human POSTN produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI6A9 | |
| Unconjugated | |
| POSTN | |
| OSF-2, OSF2, PDLPOSTN, PN | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10631 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction