missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Periostin/OSF-2 Antibody Pair [Biotin]

Product Code. 18603260 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18603260

Brand: Novus Biologicals™ NBP278863

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pair [Biotin]

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 78 to 5000 pg/mL
Assay Sensitivity 30 pg/mL
Conjugate Biotin
Product Type Sandwich ELISA
Sample Type Cell Culture Supernatant, Serum
For Use With (Application) Sandwich ELISA
Gene ID (Entrez) 10631
Gene Symbol POSTN
Quantity 1 Set
Research Discipline Cardiovascular Biology, Extracellular Matrix
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Synonym MGC119510, MGC119511, OSF-2 osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.