missing translation for 'onlineSavingsMsg'
Learn More

Periostin/OSF-2 Antibody (S03-7E3), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 444.00 - € 680.00

Specifications

Antigen Periostin/OSF-2
Clone S03-7E3
Concentration 0.3 mg/mL
Dilution Western Blot, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18642294
View Documents
Novus Biologicals
NBP3-20049-100ul
100 μg
€ 680.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18643084
View Documents
Novus Biologicals
NBP3-20049-50ul
50 μg
€ 444.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Periostin/OSF-2 Monoclonal antibody specifically detects Periostin/OSF-2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

Periostin/OSF-2
0.3 mg/mL
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cardiovascular Biology, Extracellular Matrix
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
10631
IgG
Affinity purified
S03-7E3
Western Blot, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Human, Rat
MGC119510, MGC119511, OSF-2osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1
A synthetic peptide of human Periostin/OSF-2 (Uniprot # Q15063)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.