missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peripherin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 369.00 - € 609.00
Specifications
| Antigen | Peripherin |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647205
|
Novus Biologicals
NBP2-38444-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18094824
|
Novus Biologicals
NBP2-38444 |
0.1 mL |
€ 609.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Peripherin Polyclonal specifically detects Peripherin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Peripherin | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| P41219 | |
| 5630 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: FSSTSYRRTFGPPPSLSPGAFSYSSSSRFSSSRLLGSASPSSSVRLGSFRSPRAGAGALLRLPSERLDFSMAEALNQEFLATRSNEKQEL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Neurofilament 4, neurofilament 4 (57kD), peripherin, PRPH1NEF4 | |
| PRPH | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title