missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peripherin Antibody (OTI1E10), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73337AF488
This item is not returnable.
View return policy
Description
Peripherin Monoclonal specifically detects Peripherin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Peripherin | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PRPH | |
| Full length recombinant protein of human Peripherin (NP_006253) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1E10 | |
| Western Blot, Immunohistochemistry | |
| Neurofilament 4, neurofilament 4 (57kD), peripherin, PRPH1NEF4 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 5630.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction