Learn More
Thermo Scientific™ Peristaltic Pump Tubing for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS System
Product Code. 15948486 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Peristaltic Pump Tubing
Diameter (Metric):
1.295 mm
3.175 mm
Material:
Santoprene
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Diameter (Metric) Material Unit Size
15948486 Peristaltic Pump Tubing 1.295 mm Santoprene Each
16533192 Peristaltic Pump Tubing 3.175 mm Santoprene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15948486 Supplier Thermo Scientific™ Supplier No. 1320110
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Designed for durability and fitness for purpose, a range of Thermo Scientific™ Peristaltic Pump Tubing for the iCAP™ MX and iCAP™ Q/Qnova Series is available to suit your sample matrices and ICP-MS applications.
Suit your sample matrices and applications with this comprehensive range of Thermo Scientific™ Peristaltic Pump Tubing for the Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems; all designed for durability and fitness for purpose.
Select High Quality Consumables for Improved Performance
Choose from the following Peristaltic Pump Tubing:
  • Peristaltic Pump Tubing - dia.0,508 -Orange/Yellow -PVC
  • Peristaltic Pump Tubing - dia.0,381 -Orange/Green - PVC
  • Peristaltic Pump Tubing - dia.0,762 - Black/Black -PVC
  • Peristaltic Pump Tubing - dia.0,254 - Orange/Blue -PVC
  • Peristaltic Pump Tubing - dia.1,295 - Gray/Gray -Santoprene
  • Peristaltic Pump Tubing - dia.3,175 - Black/White -Santoprene
Specifications
Type Peristaltic Pump Tubing
For Use With (Equipment) iCAP MX and iCAP Q/Qnova Series
Material Santoprene
Diameter (Metric) 1.295 mm