Learn More
Macherey-Nagel™ Permabond™ SE-52 GC Capillary Column
Product Code. 10717472 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric) Inner:
0.25 mm
0.32 mm
Film Thickness:
0.25 μm
0.5 μm
Unit Size:
Each
Product Code. 10717472
Brand: Macherey-Nagel™ 723310.25
Only null left
Specifications
Diameter (Metric) 0.5 mm
Type GC Capillary Column
Diameter (Metric) Inner 0.32 mm
Stationary Phase Permabond™ SE-52
USP Type G27, G36
For Use With (Application) Gas Chromatography (GC)
Temperature Range 300°C to 320°C
Film Thickness 0.25 μm
Length (Metric) 25 m
Packing Material Permabond™ SE-52
Quantity 1 Each