Learn More
Brady™ Permanent Self-laminating Vinyl Wrap Around Wire and Cable Labels for M6/M7 Printers
Product Code. 17831649
Click to view available options
Dimensions (LxW):
0.25 x 1.1 in.
1 x 1.9 in.
Height (Metric):
6.35 mm
25.4 mm
Unit Size:
Pack of 250
Pack of 750
Product Code. 17831649
Brand: Brady™ 173695
Only null left
Add to basket
Add to basket
Self-laminating Vinyl is an ideal material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing.
  • Self-laminating vinyl is ideal for use in cable and wire labeling and general-purpose marking in labs
  • Durable, permanent acrylic adhesive has outstanding water and oil resistance
  • UL approved
  • Service temperature of -40°C to 70°C (-40°F to 158°F)
  • Made of B-427 vinyl
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized
Adhesive Type Permanent Acrylic
Coating None
Color White
Finish Matte
Format Die-cut
Material Vinyl
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling, Wire and Cable Labeling
For Use With (Equipment) M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
Latex Free No
Quantity 750 Labels
Shape Rectangle
Temperature Range (English) -94°F to +158°F
Temperature Range (Metric) -70°C to +70°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Dimensions (LxW) 0.25 x 1.1 in.
Length (English) 0.25 in.
Width (English) 1.1 in.
Width (Metric) 27.94 mm
Height (Metric) 6.35 mm
Alternate Part Number M6-105-427-AW