missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peroxiredoxin 2 Antibody (SAIC-34D-11), Alexa Fluor™ 594 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20103AF594
This item is not returnable.
View return policy
Description
Peroxiredoxin 2 Monoclonal antibody specifically detects Peroxiredoxin 2 in Human samples. It is validated for Western Blot
Specifications
| Peroxiredoxin 2 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| EC 1.11.1, MGC4104, Natural killer cell-enhancing factor B, natural killer-enhancing factor B, NKEFBNKEF-B, peroxiredoxin 2, peroxiredoxin-2, PRPTDPX1, PRX2, PRXII, thiol-specific antioxidant 1, Thiol-specific antioxidant protein, Thioredoxin peroxidase 1, Thioredoxin-dependent peroxide reductase 1, torin, TPX1, TSAEC 1.11.1.15 | |
| Peptide "e;DHPFGFVAVPTK"e;, derived from the cellular redox regulator Peroxiredoxin-2, conjugated to KLH. | |
| 0.1 mL | |
| Cancer | |
| 7001 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot | |
| SAIC-34D-11 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction