missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peroxiredoxin 3 Antibody (AT1F8), DyLight 488 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-12869G
This item is not returnable.
View return policy
Description
Peroxiredoxin 3 Monoclonal antibody specifically detects Peroxiredoxin 3 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Peroxiredoxin 3 | |
| Monoclonal | |
| DyLight 488 | |
| Antioxidant protein 1MGC104387, AOP-1MGC24293, AOP1PRO1748, EC 1.11.1.15, HBC189, MER5, peroxiredoxin 3, Peroxiredoxin III, peroxiredoxin-3, Protein MER5 homolog, prx-III, SP-22, thioredoxin-dependent peroxide reductase, mitochondrial | |
| Recombinant human Peroxiredoxin3 (63-256aa) purified from E. coli (Accession number: NP_006784) | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 10935 | |
| Store at 4°C in the dark. | |
| IgG3 κ |
| Western Blot, ELISA, Immunofluorescence | |
| AT1F8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction