missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peroxiredoxin 4 Antibody (OTI9A3), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73338APC
This item is not returnable.
View return policy
Description
Peroxiredoxin 4 Monoclonal antibody specifically detects Peroxiredoxin 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Peroxiredoxin 4 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI9A3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| Antioxidant enzyme AOE372, AOE37-2PRX-4, EC 1.11.1.15, peroxiredoxin 4, Peroxiredoxin IV, peroxiredoxin-4, prx4, prx-4, Prx-IV, thioredoxin peroxidase (antioxidant enzyme), Thioredoxin peroxidase AO372, Thioredoxin-dependent peroxide reductase A0372 | |
| Full length human recombinant protein of human PRDX4 (NP_006397) produced in HEK293T cell. | |
| 0.1 mL | |
| Signal Transduction | |
| 10549 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction