missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peroxiredoxin 4 Antibody (S09-7F9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19736-100ul
This item is not returnable.
View return policy
Description
Peroxiredoxin 4 Monoclonal antibody specifically detects Peroxiredoxin 4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Peroxiredoxin 4 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S09-7F9 | |
| Unconjugated | |
| Antioxidant enzyme AOE372, AOE37-2PRX-4, EC 1.11.1.15, peroxiredoxin 4, Peroxiredoxin IV, peroxiredoxin-4, prx4, prx-4, Prx-IV, thioredoxin peroxidase (antioxidant enzyme), Thioredoxin peroxidase AO372, Thioredoxin-dependent peroxide reductase A0372 | |
| A synthetic peptide of human Peroxiredoxin 4 (Uniprot # Q13162) | |
| 100 μg | |
| Signal Transduction | |
| 10549 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction