missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peroxiredoxin 4 Mouse anti-Human, Clone: CPTC-PRDX4-2, Novus Biologicals™
Mouse Monoclonal Antibody
€ 484.00
Specifications
| Antigen | Peroxiredoxin 4 |
|---|---|
| Clone | CPTC-PRDX4-2 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA |
| Applications | ELISA |
Description
Peroxiredoxin 4 Monoclonal specifically detects Peroxiredoxin 4 in Human samples. It is validated for ELISA.Specifications
| Peroxiredoxin 4 | |
| 0.2 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 10549 | |
| IgG2b κ | |
| Protein A or G purified |
| CPTC-PRDX4-2 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Antioxidant enzyme AOE372, AOE37-2PRX-4, EC 1.11.1.15, peroxiredoxin 4, Peroxiredoxin IV, peroxiredoxin-4, prx4, prx-4, Prx-IV, thioredoxin peroxidase (antioxidant enzyme), Thioredoxin peroxidase AO372, Thioredoxin-dependent peroxide reductase A0372 | |
| Recombinant full-length human Peroxiredoxin 4 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title