missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PERP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110879
This item is not returnable.
View return policy
Description
Immunogen sequence: LQSSDHGQTS SLWWKCSQEG GGSGSYEEGC QSLMEYAW Highest antigen sequence indentity to the following orthologs: Rat - 82%, Mouse - 82%.
The p53 tumor-suppressor gene integrates numerous signals that control cell life and death. Several novel molecules involved in p53 network, including Chk2, p53R2, p53AIP1, Noxa, PIDD, PID/MTA2, MTBP and PERP, were identified and their genes were cloned recently. PERP, also termed PIGPC1 and THW, is a plasma membrane protein. p53 binds to the promoter of PERP and transcriptionally activates PERP gene then the translated PERP protein mediates the p53 induced apoptosis. The expression of PERP causes cell death. PERP is a mediator of p53 induced apoptosis. PERP has sequence similarity to PMP-22/gas3 and is a new member of the PMP-22/gas3 family.
Specifications
| PERP | |
| Polyclonal | |
| Unconjugated | |
| PERP | |
| 1110017A08Rik; dJ496H19.1; HGNC:17637; KCP1; KCP-1; keratinocyte associated protein 1; keratinocyte-associated protein 1; keratinocytes associated protein 1; KRTCAP1; p53 apoptosis effector related to PMP22; p53 apoptosis effector related to PMP-22; p53 apoptosis-associated target; p53-induced protein PIGPC1; PERP; PERP, TP53 apoptosis effector; PIGPC1; RP3-496H19.1; THW; Transmembrane protein THW | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64065 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96FX8 | |
| PERP | |
| Recombinant Protein corresonding to Human PERP. Recombinant protein control fragment (Product #RP-88749). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction