missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ PETG Sterile 125 mL Media Bottle without Closure
Description
Pack and ship liquid medias, buffers and sera using Thermo Scientific™ Nalgene PETG Square Media Bottles. Heavy-walled, durable bottles are designed for use in filling lines; closures sold separately.
Features:
- Heavy-walled and durable
- Shatter-resistant
- Excellent gas barrier properties
- Sterile 10-6 SAL
- Molded in graduations
- Closure supplied separately
Specifications
Specifications
| Capacity (Metric) | 125 mL |
| Diameter (Metric) Inner Neck | 28 mm |
| For Use With (Application) | Filling, Storage, Shipping |
| Material | PETG |
| No. per Case | 96 |
| Sterility | Sterile |
| Closure Included | No |
| Graduated | Yes |
| Mouth | Narrow |
| Fit for Purpose | Sterile |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?