missing translation for 'onlineSavingsMsg'
Learn More

Petroleum ether, Laboratory Reagent, bp 40-60 °C, Honeywell™

Catalog Number p-7161450
Click to view available options
Quantity:
5 L
4 x 5 L
Packaging:
Aluminum bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
64742-49-0
C6H14
86.178
MFCD00081849
AFABGHUZZDYHJO-UHFFFAOYSA-N
7892
2-methylpentane
CCCC(C)C
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
64742-49-0
C6H14
86.178
MFCD00081849
AFABGHUZZDYHJO-UHFFFAOYSA-N
7892
2-methylpentane
CCCC(C)C

Laboratory Reagent, bp 40-60 °C

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.65g/cu.cm at 20°C
Boiling Point 40°C to 60°C (1013 hPa)
Flash Point −30°C (−22°F)
Odor Characteristic
Packaging Aluminum bottle
Quantity 5 L
UN Number UN1268
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane
Solubility Information Practically insoluble in water
Grade Laboratory Reagent
Physical Form Liquid
Chemical Name or Material Petroleum Ether
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'