Learn More
Petroleum ether, pure, boiling range ca. 180-280°C
Product Code. p-3212776 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Glass Bottle
Metal drum
Plastic drum
Unit Size:
1 L
2.5 L
25 L
5 L
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10720064
Brand: Thermo Scientific Acros 387490010
Request Bulk or Custom Format
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
CAS: 8008-20-6
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 8008-20-6
MDL Number MFCD00135561
Specifications
Color Colorless
Density 0.7900g/mL
Boiling Point 200.0°C to 250.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Refractive Index 1.4300 to 1.4500
Quantity 1 L
Merck Index 15,5342
Specific Gravity 0.79
Solubility Information Solubility in water: insoluble
Grade Pure
Physical Form Liquid
Chemical Name or Material Petroleum ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Petroleum ether, boiling range ca. 180-280°C
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.