missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
€ 185.00 - € 393.00
Specifications
| Antigen | PEX1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17999142
|
Invitrogen™
PA5145450 |
20 μL |
€ 185.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17979892
|
Invitrogen™
PA5145449 |
200 μL |
€ 393.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the AAA ATPase family, a large group of ATPases associated with diverse cellular activities. This protein is cytoplasmic but is often anchored to a peroxisomal membrane where it forms a heteromeric complex and plays a role in the import of proteins into peroxisomes and peroxisome biogenesis. Mutations in this gene have been associated with complementation group 1 peroxisomal disorders such as neonatal adrenoleukodystrophy, infantile Refsum disease, and Zellweger syndrome. Alternatively spliced transcript variants have been found for this gene.Specifications
| PEX1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Pig | |
| O43933, Q5BL07 | |
| 100626142, 5189, 71382 | |
| Recombinant Human PEX1 protein, Glu62-Gln219 (Accession #O43933). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 5430414H02Rik; E330005K07Rik; HMLR1; PBD1A; PBD1B; Peroxin-1; peroxisomal biogenesis factor 1; peroxisome biogenesis disorder protein 1; Peroxisome biogenesis factor 1; Pex1; RGD1559939; Zellweger syndrome; ZWS; ZWS1 | |
| Pex1 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Pex1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title