Learn More
Invitrogen™ PEX11A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568263
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
PEX11A may be involved in peroxisomal proliferation and may regulate peroxisomes division. It may mediate binding of coatomer proteins to the peroxisomal membrane.
Specifications
| PEX11A | |
| Polyclonal | |
| Unconjugated | |
| PEX11A | |
| 28 kDa peroxisomal integral membrane protein; hsPEX11p; Peroxin-11A; peroxisomal biogenesis factor 11 alpha; peroxisomal biogenesis factor 11a; Peroxisomal coatomer receptor; peroxisomal membrane protein 11A; Peroxisomal membrane protein 26; peroxisomal membrane protein Pmp26p (Peroxin-11); Pex11; Pex11a; PEX11alpha; PEX11-alpha; pex11palpha; Pmp26p; PMP28; protein PEX11 homolog alpha; RnPEX11p | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18631, 85249, 8800 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O70597, O75192, Q9Z211 | |
| PEX11A | |
| Synthesized peptide derived from human PEX11A. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |