missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49599
This item is not returnable.
View return policy
Description
PEX12 Polyclonal antibody specifically detects PEX12 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PEX12 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| PAF3, PAF-3, peroxin 12, Peroxin-12, peroxisomal biogenesis factor 12, Peroxisome assembly factor 3, peroxisome assembly protein 12 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KAQHHSPLLRLAGVQLGRLTVQDIQALEHKPAKASMMQQPARSVSEKINSAL | |
| 0.1 mL | |
| Signal Transduction | |
| 5193 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction