missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEX12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569205
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Peroxins (PEXs) are proteins that are essential for the assembly of functional peroxisomes. The peroxisome biogenesis disorders (PBDs) are a group of genetically heterogeneous autosomal recessive, lethal diseases characterized by multiple defects in peroxisome function. The peroxisomal biogenesis disorders are a heterogeneous group with at least 14 complementation groups and with more than 1 phenotype being observed in cases falling into particular complementation groups. Although the clinical features of PBD patients vary, cells from all PBD patients exhibit a defect in the import of one or more classes of peroxisomal matrix proteins into the organelle.
Specifications
| PEX12 | |
| Polyclonal | |
| Unconjugated | |
| PEX12 | |
| AI451906; PAF3; PAF-3; PBD3A; peroxin 12; peroxin-12; peroxisomal biogenesis factor 12; peroxisome assembly factor 3; Peroxisome assembly protein 12; Pex12 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 116718 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O88177 | |
| PEX12 | |
| synthetic peptide directed towards the C-terminal region of rat PEX12. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction