missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX14 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71841UV
This item is not returnable.
View return policy
Description
PEX14 Polyclonal antibody specifically detects PEX14 in Human samples. It is validated for Western Blot
Specifications
| PEX14 | |
| Polyclonal | |
| Western Blot | |
| dJ734G22.2, MGC12767, NAPP2, NF-E2 associated polypeptide 2, peroxin-14, peroxisomal biogenesis factor 14, Peroxisomal membrane anchor protein PEX14, peroxisomal membrane anchor protein Pex14p, peroxisomal membrane protein PEX14, Pex14p, PTS1 receptor docking protein, PTS1 receptor-docking protein | |
| The immunogen for this product maps to a region between residue 250 and 300 of human Peroxisomal Biogenesis Factor 14 using the numbering given in entry NP_004556.1 (GeneID 5195). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 5195 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction