missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX5 Monoclonal Antibody (OTI6G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501406S
This item is not returnable.
View return policy
Description
PEX5 Monoclonal antibody specifically detects PEX5 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| PEX5 | |
| Monoclonal | |
| 0.87 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UGB7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55586 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunocytochemistry, Western Blot | |
| OTI6G8 | |
| Unconjugated | |
| MIOX | |
| AW212715; ESTM1; LOW QUALITY PROTEIN: peroxisomal biogenesis factor 5; PBD2A; PBD2B; peroxin 5; peroxin-5; peroxisomal biogenesis factor 5; peroxisomal C-terminal targeting signal import receptor; peroxisomal targeting signal 1 (SKL type) receptor; peroxisomal targeting signal 1 receptor; peroxisomal targeting signal 1 receptor; peroxisomal biogenesis factor 5; peroxisomal targeting signal import receptor; peroxisomal targeting signal receptor 1; peroxisome biogenesis factor 5; peroxisome receptor 1; PEX5; Pex5_predicted; PTS1 BP; PTS1 receptor; PTS1-BP; PTS1R; Pxr1; PXR1P; RCDP5; X83306 | |
| Full length human recombinant protein of human MIOX produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction