missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-43758
This item is not returnable.
View return policy
Description
Specifically detects PEX6 in Human samples, and it is validated for Western Blotting
PEX6 Polyclonal specifically detects PEX6 in Human samples. It is validated for Western Blot.
Specifications
| PEX6 | |
| Polyclonal | |
| Western Blot 1:500-1:3000 | |
| PAF2, PAF-2peroxisome assembly factor 2, Peroxin-6, peroxisomal biogenesis factor 6peroxin-6, Peroxisomal-type ATPase 1, peroxisome biogenesis factor 6, PXAAA1peroxisomal AAA-type ATPase 1 | |
| Rabbit | |
| 104 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| PEX6 | |
| Recombinant protein encompassing a sequence within the center region of human PEX6. The exact sequence is proprietary. | |
| Immunogen affinity purified | |
| RUO | |
| 5190 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction