Learn More
PF-01247324, MedChemExpress
Product Code. 16493338
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493338 10 mg 10mg
16466067 5 mg 5mg
16462726 25 mg 25mg
16454586 100 mg 100mg
16449876 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16493338 Supplier Medchem Express Supplier No. HY101383
MedChemExpress PF-01247324 is a selective and orally bioavailable Nav1.8 channel blocker with an IC50 of 196 nM for recombinant human Nav1.8 channel.
PF-01247324 is a selective and orally bioavailable Nav1.8 channel blocker with an IC50 of 196 nM for recombinant human Nav1.8 channel.
Specifications
Chemical Name or Material PF-01247324
CAS 875051-72-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C13H10Cl3N3O
Quantity 10 mg
Solubility Information DMSO : ≥ 30 mg/mL (90.74 mM)
SMILES O=C(C1=NC(N)=C(C2=CC(Cl)=CC(Cl)=C2Cl)C=C1)NC
Molecular Weight (g/mol) 330.6
Formula Weight 330.6
Percent Purity 99.42%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only