missing translation for 'onlineSavingsMsg'
Learn More

PF-06459988, MedChemExpress

Product Code. 16497998
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497998

Brand: Medchem Express HY19985

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-06459988 is an orally activity, irreversible and mutant-selective inhibitor of EGFR mutant forms. PF-06459988 demonstrates high potency and specificity to the T790M-containing double mutant EGFRs. PF-06459988 can be used for the research of cancer.

PF-06459988 is an orally activity, irreversible and mutant-selective inhibitor of EGFR mutant forms. PF-06459988 demonstrates high potency and specificity to the T790M-containing double mutant EGFRs. PF-06459988 can be used for the research of cancer.

Specifications

Chemical Name or Material PF-06459988
Physical Form Solid
CAS 1428774-45-1
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H22ClN7O3
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 50 mg/mL (115.77 mM; Need ultrasonic)
SMILES C=CC(N1C[C@H](COC2=C3C(NC=C3Cl)=NC(NC4=CN(C)N=C4)=N2)[C@@H](OC)C1)=O
Molecular Weight (g/mol) 431.88
Formula Weight 431.88
Percent Purity 99.49%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.