missing translation for 'onlineSavingsMsg'
Learn More

PF-06471553, MedChemExpress

Product Code. 16444188
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16444188

Brand: Medchem Express HY108339

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-06471553 is a potent, selective and orally available monoacylglycerol acyltransferase 3 (MGAT3) inhibitor, with an IC50 of 92 nM.

PF-06471553 is a potent, selective and orally available monoacylglycerol acyltransferase 3 (MGAT3) inhibitor, with an IC50 of 92 nM.

Specifications

Chemical Name or Material PF-06471553
CAS 1808094-07-6
Color White
Purity Grade Notes Research
Molecular Formula C23H25N5O4S
Quantity 10 mg
Solubility Information DMSO : ≥ 150 mg/mL (320.83 mM)
SMILES O=S(C1=CC2=C(CN(C(CC3=CC=CC(OC)=C3)=O)C2)C=C1)(NC4=NN(C5CCC5)N=C4)=O
Molecular Weight (g/mol) 467.54
Formula Weight 467.54
Percent Purity 98.18%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.