Learn More
PF 06650833, Tocris Bioscience™
Potent and selective IRAK4 inhibitor
Brand:  Tocris Bioscience 6373/5
Encompass
Product Code. 15993261
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
PF 06650833 is a potent and selective IRAK4 inhibitor (IC50 = 2 nM). Inhibits LPS-induced TNF-α secretion in vivo. Orally bioavailable.
Specifications
Specifications
1-[[(2S,3S,4S)-3-Ethyl-4-fluoro-5-oxo-2-pyrrolidinyl]methoxy]-7-methoxy-6-isoquinolinecarboxamide
1817626-54-2
>98%
5 mg