missing translation for 'onlineSavingsMsg'
Learn More

PF-5274857, MedChemExpress

Product Code. 16461857
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461857 50 mg 50mg
16498387 5 mg 5mg
16496186 100 mg 100mg
16445698 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16461857 Supplier Medchem Express Supplier No. HY13459

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-5274857 is a potent, selective, orally active and brain-penetrant antagonist of Smo, with an IC50 of 5.8 nM and Ki of 4.6 nM. PF-5274857 has potential for research of tumor types including brain tumors and brain metastasis driven by an activated Hh pathway.

PF-5274857 is a potent, selective, orally active and brain-penetrant antagonist of Smo, with an IC50 of 5.8 nM and Ki of 4.6 nM. PF-5274857 has potential for research of tumor types including brain tumors and brain metastasis driven by an activated Hh pathway.

Specifications

Chemical Name or Material PF-5274857
CAS 1373615-35-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H25ClN4O3S
Quantity 50 mg
Solubility Information DMSO : 125 mg/mL (286.07 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(N1CCN(C2=NC=C(Cl)C(C3=NC=C(C)C=C3C)=C2)CC1)CCS(=O)(C)=O
Molecular Weight (g/mol) 436.96
Formula Weight 436.96
Percent Purity 98.89%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.