missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PF-5274857, MedChemExpress
Description
PF-5274857 is a potent, selective, orally active and brain-penetrant antagonist of Smo, with an IC50 of 5.8 nM and Ki of 4.6 nM. PF-5274857 has potential for research of tumor types including brain tumors and brain metastasis driven by an activated Hh pathway.
Specifications
Specifications
| Chemical Name or Material | PF-5274857 |
| CAS | 1373615-35-0 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C20H25ClN4O3S |
| Quantity | 5 mg |
| Solubility Information | DMSO : 125 mg/mL (286.07 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | O=C(N1CCN(C2=NC=C(Cl)C(C3=NC=C(C)C=C3C)=C2)CC1)CCS(=O)(C)=O |
| Molecular Weight (g/mol) | 436.96 |
| Formula Weight | 436.96 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?