missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PF1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-81671AF532
This item is not returnable.
View return policy
Description
PF1 Polyclonal antibody specifically detects PF1 in Human, Mouse, Chicken samples. It is validated for Western Blot
Specifications
| PF1 | |
| Polyclonal | |
| Western Blot | |
| FLJ34122, KIAA1523MGC131914, PF1, PHD factor 1, PHD finger protein 12, PHD zinc finger transcription factor | |
| The immunogen recognized by this antibody maps to a region between residue 954 and 1004 of human PHD factor 1 using the numbering given in entry NP_001028733.1 (GeneID 57649). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Chicken | |
| Purified |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 57649 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction