missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00
Specifications
| Antigen | PF1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
PF1 Polyclonal specifically detects PF1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| PF1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 57649 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GSSKRRRKEETTGKNVKKTQHELDHNGLVPLPVKVCFTCNRSCRVAPLIQCDYCPLLFHMDCLEPPLTAMPLGRWMCPNHIEHVVLNQKNMTLS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| FLJ34122, KIAA1523MGC131914, PF1, PHD factor 1, PHD finger protein 12, PHD zinc finger transcription factor | |
| PHF12 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title