missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PF1 Antibody [PE], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB100-81671PE
This item is not returnable.
View return policy
Description
PF1 Polyclonal antibody specifically detects PF1 in Chicken,Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot,ChIP assay (ChIP)
Specifications
| PF1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 57649 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunoprecipitation, Western Blot, ChIP Assay | |
| PE | |
| FLJ34122, KIAA1523MGC131914, PF1, PHD factor 1, PHD finger protein 12, PHD zinc finger transcription factor | |
| The immunogen recognized by this antibody maps to a region between residue 954 and 1004 of human PHD factor 1 using the numbering given in entry NP_001028733.1 (GeneID 57649). | |
| 0.1 mL | |
| Primary | |
| Chicken, Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction