missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PF4 Monoclonal Antibody (C4)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543756
This item is not returnable.
View return policy
Description
PF4 Monoclonal Antibody for Western Blot, IHC (P)
Platelet factor-4 is a 70-amino acid protein that is released from the alpha-granules of activated platelets and binds with high affinity to heparin. Its major physiologic role appears to be neutralization of heparin-like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. As a strong chemoattractant for neutrophils and fibroblasts, PF4 probably has a role in inflammation and wound repair (Eisman et al., 1990).
Specifications
| PF4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P02776, P06765 | |
| Pf4 | |
| Recombinant protein Platelet Factor 4. The antigen corresponds to amino acid range 31-101 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Rat, Pig | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| C4 | |
| Unconjugated | |
| Pf4 | |
| chemokine (C-X-C motif) ligand 4; C-X-C motif chemokine 4; Cxcl4; Endothelial cell growth inhibitor; Iroplact; oncostatin-A; PF4; PF-4; Pf4a; Platelet factor 4; Platelet factor 4, short form; RATPF4A; Scyb4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 360918, 5196 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction