Learn More
Invitrogen™ PF4 Monoclonal Antibody (KKO)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA517641
Product Code. 15217826
  • € 524.00 / 200µg
Description
MA5-17641 targets Platelet Factor 4 in ELISA and WB applications and shows reactivity with Human samples. The MA5-17641 immunogen is human Factor IV purified from the supernatant of thrombin activated platelets.
Platelet factor-4 is a 70-amino acid protein that is released from the alpha-granules of activated platelets and binds with high affinity to heparin. Its major physiologic role appears to be neutralization of heparin-like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. As a strong chemoattractant for neutrophils and fibroblasts, PF4 probably has a role in inflammation and wound repair (Eisman et al., 1990).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PF4
Monoclonal
1 mg/mL
PBS with 0.02% sodium azide
P02776
Pf4
Human Factor IV purified from the supernatant of thrombin activated platelets.
200 μg
Primary
Human
Antibody
IgG2b κ
ELISA, Western Blot
KKO
Unconjugated
Pf4
chemokine (C-X-C motif) ligand 4; C-X-C motif chemokine 4; Cxcl4; Endothelial cell growth inhibitor; Iroplact; oncostatin-A; PF4; PF-4; Pf4a; Platelet factor 4; Platelet factor 4, short form; RATPF4A; Scyb4
Mouse
Protein G
RUO
5196
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.