Learn More
Cole-Parmer™ PFA Female Threaded Tee Fitting
Click to view available options
Thread Size:
1/2 in.
3/4 in.
3/8 in.
Unit Size:
Each
Description
- Chemically-inert PFA handles most aggressive chemicals
- Threaded reducing bushing
- Choose the size and material that meets your application needs
- Use with our rigid tubing for your high-pressure applications
Specifications
Specifications
| Description | NPT(F) Connection |
| Autoclavable | Not Autoclavable |
| Material | PFA |
| Tubing Size | 1/2 in. |
| Outlet Port | 1/2 in. |
| Port Size | 1/2 in. |
| Sterilization Method | Ethylene Oxide |
| Thread Size | 1/2 in. |
| Shape | T-shape |
| Color | Translucent |
| Show More |